Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcoln3
Entrez Gene ID
171166
Gene Name
mucolipin 3
Gene Aliases
6720490O21Rik, TRPML3, Va
Chromosome Location
Chr.3: 146093277 - 146141808 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134160.1 NP_598921.1 11 - 12 1532 97
GenBank mRNA AF475086.1 - 11 - 12 1532 97
AK030819.1 - 11 - 12 1652 97
AK033008.1 - 11 - 12 1646 97
AK035029.1 - 11 - 12 1653 97
AY083531.1 - 11 - 12 1530 97
BC106856.2 - 11 - 12 1532 97
BC106857.2 - 11 - 12 1532 97

View Full Product Details