The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011082.3 | NP_035212.2 | 10 - 11 | 2401 | 62 |
GenBank mRNA | AK128944.1 | - | 10 - 11 | 2401 | 62 |
AK142656.1 | - | 10 - 11 | 2403 | 62 | |
BC013556.1 | - | 10 - 11 | 2334 | 62 | |
U06431.1 | - | 10 - 11 | 2305 | 62 |