The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008250.2 | NP_032276.1 | 1 - 2 | 935 | 82 |
GenBank mRNA | AK154090.1 | - | 1 - 2 | 613 | 82 |
BB664380.1 | - | 1 - 2 | 559 | 82 | |
BC050146.1 | - | 1 - 2 | 190 | 82 | |
BC064068.1 | - | 1 - 2 | 935 | 82 | |
X58250.1 | - | 1 - 2 | 1051 | 82 |