Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc44a4
Entrez Gene ID
70129
Gene Name
solute carrier family 44, member 4
Gene Aliases
2210409B01Rik, NG22
Chromosome Location
Chr.17: 34914224 - 34930436 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023557.3 NP_076046.1 14 - 15 1497 55
GenBank mRNA AK008864.1 - 3 - 4 459 55
AK145621.1 - 14 - 15 1489 55
AK164684.1 - 13 - 14 2265 55
AK166620.1 - 11 - 12 1399 55
BC010808.1 - 14 - 15 1495 55
BC052652.1 - 14 - 15 1495 55

View Full Product Details