Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hnmt
Entrez Gene ID
140483
Gene Name
histamine N-methyltransferase
Gene Aliases
1500031F01Rik, AI788969
Chromosome Location
Chr.2: 24002911 - 24049757 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080462.2 NP_536710.1 1 - 2 60 64
GenBank mRNA AB070524.1 - 1 - 2 56 64
AF297037.1 - 1 - 2 60 64
BC033928.1 - 1 - 2 52 64

View Full Product Details