The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011258.2 | NP_035388.2 | 20 - 21 | 2757 | 75 |
GenBank mRNA | AK134660.1 | - | 5 - 6 | 535 | 75 |
AK143077.1 | - | 1 - 2 | 1289 | 75 | |
BC085173.1 | - | 20 - 21 | 2734 | 75 | |
BC085174.1 | - | 20 - 21 | 2724 | 75 | |
U01222.1 | - | 20 - 21 | 2725 | 75 | |
U07157.1 | - | 20 - 21 | 2720 | 75 | |
U36441.1 | - | 20 - 21 | 2731 | 75 | |
X72711.1 | - | 20 - 21 | 2723 | 75 |