Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Brix1
Entrez Gene ID
67832
Gene Name
BRX1, biogenesis of ribosomes, homolog (S. cerevisiae)
Gene Aliases
1110064N10Rik, Bxdc2, C76935
Chromosome Location
Chr.15: 10474779 - 10485937 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026396.3 NP_080672.3 7 - 8 601 71
GenBank mRNA AK002985.1 - 7 - 8 488 71

View Full Product Details