The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029612.4 | NP_083888.3 | 2 - 3 | 519 | 93 |
GenBank mRNA | AB196824.1 | - | 2 - 3 | 382 | 93 |
AB196825.1 | - | 2 - 3 | 382 | 93 | |
AB196826.1 | - | 2 - 3 | 382 | 93 | |
AB196827.1 | - | 2 - 3 | 382 | 93 | |
AK009505.1 | - | 2 - 3 | 475 | 93 | |
AK090041.1 | - | 2 - 3 | 510 | 93 | |
BC019477.1 | - | 2 - 3 | 458 | 93 |