The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033561.1 | NP_291039.1 | 6 - 7 | 637 | 88 |
GenBank mRNA | AK045004.1 | - | 6 - 7 | 639 | 88 |
AK088818.1 | - | 6 - 7 | 649 | 88 | |
AK122194.1 | - | 4 - 5 | 3859 | 88 | |
AK132034.1 | - | 6 - 7 | 657 | 88 | |
AK135390.1 | - | 6 - 7 | 638 | 88 | |
AK136999.1 | - | 5 - 6 | 564 | 88 | |
AK149841.1 | - | 6 - 7 | 623 | 88 | |
AK150088.1 | - | 6 - 7 | 646 | 88 | |
AK166567.1 | - | 6 - 7 | 638 | 88 | |
AK168872.1 | - | 5 - 6 | 596 | 88 | |
AK169312.1 | - | 6 - 7 | 620 | 88 | |
AK169353.1 | - | 6 - 7 | 638 | 88 | |
AK169407.1 | - | 5 - 6 | 596 | 88 | |
AK188319.1 | - | 2 - 3 | 233 | 88 | |
AK210675.1 | - | 1 - 2 | 89 | 88 | |
AK219562.1 | - | 1 - 2 | 89 | 88 | |
BC014796.1 | - | 5 - 6 | 591 | 88 |