The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026582.4 | NP_080858.3 | 3 - 4 | 753 | 118 |
NR_037590.1 | 4 - 5 | 834 | 118 | ||
GenBank mRNA | AK019886.1 | - | 4 - 5 | 792 | 118 |
AK029053.1 | - | 3 - 4 | 712 | 118 | |
AK029104.1 | - | 3 - 4 | 710 | 118 | |
AK029315.1 | - | 3 - 4 | 710 | 118 | |
AK031444.1 | - | 3 - 4 | 710 | 118 | |
AK044810.1 | - | 3 - 4 | 712 | 118 | |
AK049842.1 | - | 3 - 4 | 710 | 118 | |
AK077407.1 | - | 3 - 4 | 711 | 118 | |
AK128912.1 | - | 3 - 4 | 709 | 118 | |
AK159207.1 | - | 3 - 4 | 710 | 118 | |
AK159977.1 | - | 3 - 4 | 712 | 118 | |
BC018381.1 | - | 2 - 3 | 337 | 118 | |
BC075615.1 | - | 3 - 4 | 753 | 118 |