The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030113.2 | NP_084389.2 | 20 - 21 | 2156 | 103 |
GenBank mRNA | AF297029.1 | - | 20 - 21 | 2060 | 103 |
AF297030.1 | - | 20 - 21 | 2119 | 103 | |
AY179965.1 | - | 20 - 21 | 2032 | 103 | |
AY179966.1 | - | 19 - 20 | 1966 | 103 | |
CF106470.1 | - | 3 - 4 | 433 | 103 |