Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ltb4r1
Entrez Gene ID
16995
Gene Name
leukotriene B4 receptor 1
Gene Aliases
BLT1, BLTR, Ltb4r, mBLTR
Chromosome Location
Chr.14: 55765050 - 55768494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008519.2 NP_032545.1 1 - 2 134 76
GenBank mRNA AF044030.1 - 1 - 2 130 76
AF110104.1 - 1 - 2 81 76
AK010895.1 - 1 - 2 119 76
AK155181.1 - 1 - 2 135 76
BC064063.1 - 1 - 2 65 76

View Full Product Details