The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145572.2 | NP_663547.2 | 8 - 9 | 1478 | 97 |
GenBank mRNA | AK138992.1 | - | 8 - 9 | 1186 | 97 |
AK149391.1 | - | 8 - 9 | 1202 | 97 | |
BC021322.1 | - | 8 - 9 | 1339 | 97 | |
BC158081.1 | - | 8 - 9 | 1339 | 97 |