Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm5a
Entrez Gene ID
214899
Gene Name
lysine (K)-specific demethylase 5A
Gene Aliases
AA409370, C76986, Jarid1a, RBP2, Rbbp2
Chromosome Location
Chr.6: 120364099 - 120444574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145997.2 NP_666109.2 14 - 15 2486 124
GenBank mRNA AK144877.1 - 14 - 15 2457 124
AK159079.1 - 14 - 15 2463 124
AK166055.1 - 14 - 15 2467 124
BC010717.1 - 14 - 15 2152 124
BC011506.3 - 14 - 15 2420 124
BC080691.1 - 14 - 15 2172 124

View Full Product Details