The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172426.2 | NP_766014.1 | 1 - 2 | 220 | 92 |
GenBank mRNA | AK034424.1 | - | 1 - 2 | 104 | 92 |
AK038634.1 | - | 1 - 2 | 104 | 92 | |
AK038800.1 | - | 1 - 2 | 148 | 92 | |
AK083127.1 | - | 1 - 2 | 104 | 92 | |
AK136058.1 | - | 1 - 2 | 148 | 92 | |
CJ121602.1 | - | 1 - 2 | 223 | 92 |