Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stradb
Entrez Gene ID
227154
Gene Name
STE20-related kinase adaptor beta
Gene Aliases
AA792893, Als2cr2, B830008M19, D1Ucla2, ILPIP, ILPIPA, PRO1038, Papk, Syradb
Chromosome Location
Chr.1: 58973527 - 58995126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172656.5 NP_766244.4 5 - 6 689 108
GenBank mRNA AB057666.1 - 5 - 6 456 108
AB057667.1 - 5 - 6 456 108
AK080096.1 - 5 - 6 653 108
AK086974.1 - 5 - 6 620 108
AK140272.1 - 5 - 6 739 108
AK146079.1 - 5 - 6 502 108
BC037003.1 - 5 - 6 598 108
BC055814.1 - 6 - 7 829 108
BC060956.1 - 5 - 6 546 108

View Full Product Details