The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168290.1 | NP_001161762.1 | 2 - 3 | 549 | 72 |
NM_172755.3 | NP_766343.3 | 2 - 3 | 549 | 72 | |
GenBank mRNA | AF518875.1 | - | 2 - 3 | 542 | 72 |
AK042293.1 | - | 2 - 3 | 550 | 72 | |
AK137294.1 | - | 2 - 3 | 153 | 72 | |
BC057305.1 | - | 2 - 3 | 203 | 72 |