Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d4
Entrez Gene ID
210789
Gene Name
TBC1 domain family, member 4
Gene Aliases
5930406J04Rik, A930035N22, AV295684, As160
Chromosome Location
Chr.14: 101442360 - 101609228 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081278.2 NP_001074747.2 4 - 5 2029 93
GenBank mRNA AK031120.1 - 4 - 5 1873 93
AK039337.1 - 4 - 5 2030 93
AK044719.1 - 4 - 5 1671 93
AK220347.1 - 3 - 4 436 93
BC042747.1 - 1 - 2 123 93
BC064433.1 - 4 - 5 1653 93
BC080762.1 - 4 - 5 1647 93
BC117869.1 - 4 - 5 1505 93
BC117870.1 - 4 - 5 1505 93

View Full Product Details