The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038846.1 | NP_001033935.1 | 1 - 2 | 205 | 74 |
NM_178593.3 | NP_848708.2 | 1 - 2 | 205 | 74 | |
GenBank mRNA | AK133961.1 | - | 1 - 2 | 160 | 74 |
AK158429.1 | - | 1 - 2 | 146 | 74 | |
AK168816.1 | - | 1 - 2 | 176 | 74 | |
AK171323.1 | - | - | 185 | 74 | |
AK171914.1 | - | 1 - 2 | 128 | 74 | |
BC025872.1 | - | 1 - 2 | 125 | 74 | |
BY230131.1 | - | 1 - 2 | 206 | 74 |