Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdca5
Entrez Gene ID
67849
Gene Name
cell division cycle associated 5
Gene Aliases
2610036L13Rik, AL024086, AW536684, C85404
Chromosome Location
Chr.19: 6085097 - 6091773 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026410.3 NP_080686.1 4 - 5 299 148
GenBank mRNA AK010540.1 - 4 - 5 266 148
AK011701.1 - 4 - 5 299 148
AK016305.1 - 4 - 5 413 148
BC029626.1 - 3 - 4 193 148
BC052904.1 - 4 - 5 297 148

View Full Product Details