Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Med16
Entrez Gene ID
216154
Gene Name
mediator complex subunit 16
Gene Aliases
95kDa, A630083L04, Thrap5, Trap95
Chromosome Location
Chr.10: 79894707 - 79908941 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163276.1 NP_001156748.1 7 - 8 1255 63
NM_198107.2 NP_932775.2 7 - 8 1258 63
GenBank mRNA AK042340.1 - 7 - 8 1220 63
AK149345.1 - 7 - 8 1239 63
BC057056.1 - 7 - 8 1212 63

View Full Product Details