Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr36
Entrez Gene ID
225348
Gene Name
WD repeat domain 36
Gene Aliases
5730444A13Rik, Ta-wdrp
Chromosome Location
Chr.18: 32837225 - 32866420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110015.1 NP_001103485.1 2 - 3 359 82
NM_001110016.1 NP_001103486.1 2 - 3 359 82
NM_144863.4 NP_659112.2 2 - 3 359 82
GenBank mRNA AK041763.1 - 2 - 3 279 82
AK045396.1 - 2 - 3 332 82
AK077545.1 - 2 - 3 289 82
AK140019.1 - 2 - 3 284 82
AK155965.1 - 2 - 3 258 82
AK156822.1 - - 360 82
AK171683.1 - 2 - 3 297 82
AK172054.1 - 2 - 3 284 82
AK172377.1 - 2 - 3 289 82
BC022132.1 - 2 - 3 331 82
BC025809.1 - 2 - 3 273 82
BC094453.1 - 2 - 3 310 82

View Full Product Details