The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199476.1 | NP_955770.1 | 8 - 9 | 1062 | 152 |
GenBank mRNA | AK138731.1 | - | 8 - 9 | 1055 | 152 |
BC058103.1 | - | 8 - 9 | 1062 | 152 |