The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172769.2 | NP_766357.1 | 3 - 4 | 401 | 109 |
GenBank mRNA | AB016248.1 | - | 3 - 4 | 387 | 109 |
AK043825.1 | - | 3 - 4 | 360 | 109 | |
AK077670.1 | - | 3 - 4 | 359 | 109 | |
AK154309.1 | - | 3 - 4 | 348 | 109 | |
BC024132.1 | - | 3 - 4 | 350 | 109 |