Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lpp
Entrez Gene ID
210126
Gene Name
LIM domain containing preferred translocation partner in lipoma
Gene Aliases
9430020K16Rik, AA959454, AU024130, B130055L10Rik, C79715, D630048H16
Chromosome Location
Chr.16: 24393350 - 24992578 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145952.1 NP_001139424.1 7 - 8 1456 99
NM_001145954.1 NP_001139426.1 3 - 4 870 99
NM_178665.5 NP_848780.3 8 - 9 1663 99
GenBank mRNA AK029567.1 - 3 - 4 871 99
AK040643.1 - 6 - 7 1217 99
AK045288.1 - 8 - 9 1524 99
AK051345.1 - 8 - 9 1535 99
AK054550.1 - 7 - 8 1459 99
AK076989.1 - 3 - 4 861 99
AK085626.1 - 8 - 9 1655 100
AK168431.1 - 8 - 9 1570 99
AK169780.1 - 8 - 9 1475 99
BC085321.1 - 7 - 8 1402 99

View Full Product Details