The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175535.3 | NP_780744.2 | 14 - 15 | 1875 | 61 |
GenBank mRNA | AK129345.1 | - | 11 - 12 | 1364 | 61 |