The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177633.4 | NP_808301.3 | 3 - 4 | 321 | 80 |
GenBank mRNA | AK147730.1 | - | 3 - 4 | 317 | 80 |