The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172788.3 | NP_766376.2 | 1 - 2 | 766 | 63 |
GenBank mRNA | AK133071.1 | - | 1 - 2 | 766 | 63 |