Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
L3mbtl3
Entrez Gene ID
237339
Gene Name
l(3)mbt-like 3 (Drosophila)
Gene Aliases
AI481284, MBT-1
Chromosome Location
Chr.10: 26274470 - 26375989 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172787.2 NP_766375.1 6 - 7 673 66
GenBank mRNA AK031398.1 - 6 - 7 659 66
AK045667.1 - 6 - 7 674 66
AK049597.1 - 6 - 7 708 66
AK129443.1 - 2 - 3 218 66
BC082309.1 - 5 - 6 543 66
BC085192.1 - 5 - 6 778 66

View Full Product Details