Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm5a
Entrez Gene ID
214899
Gene Name
lysine (K)-specific demethylase 5A
Gene Aliases
AA409370, C76986, Jarid1a, RBP2, Rbbp2
Chromosome Location
Chr.6: 120364099 - 120444574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145997.2 NP_666109.2 20 - 21 3553 70
GenBank mRNA AK134200.1 - 2 - 3 371 70
AK135085.1 - 5 - 6 881 70
AK144877.1 - 20 - 21 3524 70
AK159079.1 - 19 - 20 3415 70
AK166055.1 - 20 - 21 3534 70
BC010717.1 - 19 - 20 3104 70
BC011506.3 - 20 - 21 3487 70
BC022581.1 - 5 - 6 828 70
BC080691.1 - 20 - 21 3239 70

View Full Product Details