Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm5a
Entrez Gene ID
214899
Gene Name
lysine (K)-specific demethylase 5A
Gene Aliases
AA409370, C76986, Jarid1a, RBP2, Rbbp2
Chromosome Location
Chr.6: 120364099 - 120444574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145997.2 NP_666109.2 8 - 9 1537 84
GenBank mRNA AK144877.1 - 8 - 9 1508 84
AK159079.1 - 8 - 9 1514 84
AK166055.1 - 8 - 9 1518 84
BC010717.1 - 8 - 9 1178 84
BC011506.3 - 8 - 9 1471 84
BC080691.1 - 8 - 9 1223 84

View Full Product Details