The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016801.3 | NP_058081.2 | 9 - 10 | 792 | 76 |
GenBank mRNA | AK144399.1 | - | 6 - 7 | 765 | 76 |
BC023379.1 | - | 1 - 2 | 185 | 76 | |
BC047133.1 | - | 9 - 10 | 781 | 76 | |
BC057892.1 | - | 9 - 10 | 887 | 76 | |
BC100446.1 | - | 9 - 10 | 827 | 76 | |
D45208.2 | - | 9 - 10 | 792 | 76 |