The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029752.2 | NP_084028.1 | 2 - 3 | 433 | 73 |
GenBank mRNA | AK010810.1 | - | 2 - 3 | 376 | 73 |
AK043288.1 | - | 2 - 3 | 375 | 73 | |
AK044565.1 | - | 2 - 3 | 321 | 73 | |
AK143538.1 | - | 2 - 3 | 392 | 73 | |
BC039941.1 | - | 2 - 3 | 391 | 73 |