The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177770.1 | NP_001171241.1 | 4 - 5 | 706 | 167 |
NM_001177771.1 | NP_001171242.1 | 5 - 6 | 621 | 167 | |
NM_145543.2 | NP_663518.1 | 4 - 5 | 418 | 167 | |
GenBank mRNA | AK090092.1 | - | 4 - 5 | 392 | 167 |
AK143631.1 | - | 5 - 6 | 604 | 167 | |
AK169803.1 | - | 4 - 5 | 416 | 167 | |
AK171296.1 | - | 4 - 5 | 445 | 167 | |
BC003247.1 | - | 4 - 5 | 449 | 167 |