Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kifc2
Entrez Gene ID
16581
Gene Name
kinesin family member C2
Gene Aliases
-
Chromosome Location
Chr.15: 76659837 - 76668196 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010630.2 NP_034760.2 7 - 8 836 83
GenBank mRNA AK144304.1 - 8 - 9 995 83
D49545.1 - 7 - 8 912 83
U92949.1 - 7 - 8 826 83

View Full Product Details