Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iglon5
Entrez Gene ID
210094
Gene Name
IgLON family member 5
Gene Aliases
A230106M20Rik, Gm1431
Chromosome Location
Chr.7: 43472907 - 43490057 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164518.1 NP_001157990.1 3 - 4 394 68
GenBank mRNA AK039193.1 - 3 - 4 361 68

View Full Product Details