Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tacc1
Entrez Gene ID
320165
Gene Name
transforming, acidic coiled-coil containing protein 1
Gene Aliases
4833447E04Rik, AA960202, B230378H13Rik
Chromosome Location
Chr.8: 25154552 - 25241585 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177089.5 NP_796063.3 9 - 10 2127 91
NM_199323.3 NP_955355.1 7 - 8 903 91
GenBank mRNA AK158441.1 - 9 - 10 2134 91
AY177412.1 - 7 - 8 811 91
AY177413.1 - 9 - 10 2029 91
BC125389.1 - 7 - 8 713 91
BC145709.1 - 7 - 8 713 91

View Full Product Details