Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Parg
Entrez Gene ID
26430
Gene Name
poly (ADP-ribose) glycohydrolase
Gene Aliases
AI413217
Chromosome Location
Chr.14: 32201949 - 32298714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011960.2 NP_036090.2 15 - 16 2796 88
GenBank mRNA AF079557.1 - 15 - 16 2545 88
AK034542.1 - 8 - 9 728 88
AK036656.1 - 15 - 16 2819 88
AK088708.1 - 12 - 13 1151 88
AK164013.1 - 15 - 16 2783 88
BC050892.1 - 15 - 16 2795 88
BC053037.1 - 15 - 16 2780 88
BC059827.1 - 15 - 16 2805 88
EF382673.1 - 13 - 14 1505 88
JQ890227.1 - 12 - 13 1302 88

View Full Product Details