Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Glt8d1
Entrez Gene ID
76485
Gene Name
glycosyltransferase 8 domain containing 1
Gene Aliases
2410004H05Rik, 5430414N14Rik, AI450005
Chromosome Location
Chr.14: 31001347 - 31012441 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165930.1 NP_001159402.1 2 - 3 89 98
GenBank mRNA AK077388.1 - 2 - 3 83 98

View Full Product Details