Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmprss2
Entrez Gene ID
50528
Gene Name
transmembrane protease, serine 2
Gene Aliases
D16Ertd61e
Chromosome Location
Chr.16: 97564682 - 97659538 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015775.2 NP_056590.2 6 - 7 796 68
GenBank mRNA AF199362.1 - 6 - 7 677 68
AF243500.1 - 6 - 7 666 68
AK141350.1 - 6 - 7 771 68
AK146047.1 - 6 - 7 680 68
AK161173.1 - 6 - 7 790 68
BC038393.1 - 6 - 7 635 68
BC054348.1 - 6 - 7 796 68

View Full Product Details