Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbc1d4
Entrez Gene ID
210789
Gene Name
TBC1 domain family, member 4
Gene Aliases
5930406J04Rik, A930035N22, AV295684, As160
Chromosome Location
Chr.14: 101442360 - 101609228 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081278.2 NP_001074747.2 6 - 7 2252 62
GenBank mRNA AK031120.1 - 6 - 7 2096 62
AK039337.1 - 6 - 7 2253 62
AK044719.1 - 6 - 7 1894 62
AK220347.1 - 5 - 6 659 62
BC064433.1 - 6 - 7 1876 62
BC080762.1 - 6 - 7 1870 62
BC117870.1 - 6 - 7 1728 62

View Full Product Details