Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcnxl3
Entrez Gene ID
104401
Gene Name
pecanex-like 3 (Drosophila)
Gene Aliases
mKIAA4073
Chromosome Location
Chr.19: 5664635 - 5688908 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144868.3 NP_659117.2 9 - 10 2858 81
GenBank mRNA AF237953.1 - 7 - 8 875 81
BC025838.1 - 3 - 4 391 81
BC038056.1 - 4 - 5 966 81
BC054751.1 - 9 - 10 2137 81
BC055289.1 - 10 - 11 2661 81
BC096538.1 - 9 - 10 2858 81

View Full Product Details