Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpifb4
Entrez Gene ID
381399
Gene Name
BPI fold containing family B, member 4
Gene Aliases
Gm1006, Lplunc4
Chromosome Location
Chr.2: 153938217 - 153963852 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034875.3 NP_001030047.2 3 - 4 702 87

View Full Product Details