Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zc3h18
Entrez Gene ID
76014
Gene Name
zinc finger CCCH-type containing 18
Gene Aliases
1190001B23Rik, 5830416A07Rik, Nhn1
Chromosome Location
Chr.8: 122376614 - 122417360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029993.1 NP_001025164.1 5 - 6 1101 72
NM_001029994.1 NP_001025165.1 4 - 5 1029 72
GenBank mRNA AK031413.1 - 5 - 6 1050 72
AK131189.1 - 2 - 3 173 72
AK159881.1 - 5 - 6 1037 72
AK165989.1 - 3 - 4 380 72
BC030495.1 - 4 - 5 1009 72
BC094330.1 - 2 - 3 182 72

View Full Product Details