Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ldhd
Entrez Gene ID
52815
Gene Name
lactate dehydrogenase D
Gene Aliases
4733401P21Rik, D8Bwg1320e
Chromosome Location
Chr.8: 111626271 - 111630322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027570.3 NP_081846.3 9 - 10 1187 74
GenBank mRNA AK014649.1 - 9 - 10 1193 74
AK029820.1 - - 1601 74
AK037996.1 - - 497 74
AY092768.1 - 9 - 10 1192 74
BC039155.1 - 9 - 10 1183 74
BC055443.1 - 9 - 10 1187 74

View Full Product Details