Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acta1
Entrez Gene ID
11459
Gene Name
actin, alpha 1, skeletal muscle
Gene Aliases
AA959943, Acta-2, Acts, Actsk-1
Chromosome Location
Chr.8: 123891758 - 123894775 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009606.3 NP_033736.1 1 - 2 83 97

View Full Product Details