The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029342.4 | NP_083618.3 | - | 345 | 75 |
GenBank mRNA | AK006481.1 | - | - | 339 | 75 |
BC006063.1 | - | - | 319 | 75 | |
BC107252.1 | - | - | 264 | 75 | |
BC107253.1 | - | - | 264 | 75 | |
BC132359.1 | - | - | 235 | 75 | |
BC132361.1 | - | - | 235 | 75 | |
BY095373.1 | - | - | 268 | 75 |