Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc24a2
Entrez Gene ID
76376
Gene Name
solute carrier family 24 (sodium/potassium/calcium exchanger), member 2
Gene Aliases
2810021B17Rik, 6330417K15Rik, AI847460, Nckx2
Chromosome Location
Chr.4: 86983124 - 87230554 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110240.1 NP_001103710.1 3 - 4 1301 86
GenBank mRNA BC115867.1 - 3 - 4 1303 86

View Full Product Details