The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010531.1 | NP_034661.1 | 5 - 6 | 496 | 59 |
GenBank mRNA | AB019505.1 | - | 3 - 4 | 345 | 59 |
AF110802.1 | - | 3 - 4 | 1723 | 59 | |
AF110803.1 | - | 4 - 5 | 423 | 59 | |
AF122907.1 | - | 5 - 6 | 496 | 59 | |
AK003370.1 | - | 5 - 6 | 512 | 59 | |
AK008452.1 | - | 4 - 5 | 427 | 59 | |
AK009721.1 | - | 4 - 5 | 423 | 59 | |
AK009877.1 | - | 1 - 2 | 63 | 59 | |
BC018332.1 | - | 4 - 5 | 406 | 59 | |
CT010164.1 | - | 3 - 4 | 345 | 59 | |
CT010334.1 | - | 3 - 4 | 345 | 59 |