The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027698.5 | NP_081974.3 | 6 - 7 | 674 | 92 |
GenBank mRNA | AK015831.1 | - | - | 584 | 92 |
AK016902.1 | - | - | 584 | 92 | |
AK029056.1 | - | 6 - 7 | 599 | 92 | |
AK135821.1 | - | 6 - 7 | 587 | 92 | |
AK137621.1 | - | 6 - 7 | 620 | 92 | |
AK141507.1 | - | 6 - 7 | 595 | 92 | |
AK160698.1 | - | 6 - 7 | 570 | 92 | |
AK165163.1 | - | 6 - 7 | 584 | 92 | |
BC052150.1 | - | - | 1301 | 92 | |
BC090961.1 | - | 6 - 7 | 576 | 92 |